missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MSL3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA528282

Product Code. 13412397

  • € 524.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T, mouse muscle. Predicted reactivity: Mouse (95%), Rat (94%), Xenopus laevis (80%), Pig (96%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a nuclear protein and has similarity to drosophila male-specific lethal-3 gene. The drosophila protein plays a critical role in a dosage-compensation pathway, which equalizes X-linked gene expression in males and females. Thus this encoded protein is thought to play a similar function in chromatin remodeling and transcriptional regulation. This gene has been found to undergo X inactivation. There are four alternatively spliced transcript variants of this gene encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MSL3
Polyclonal
Unconjugated
Msl3
AU018931; male-specific lethal 3 homolog; male-specific lethal 3 homolog (Drosophila); male-specific lethal-3 homolog 1; Male-specific lethal-3 protein-like 1; MSL3; Msl31; MSL3L1; MSL3-like 1
Rabbit
Antigen affinity chromatography
RUO
10943, 17692
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q8N5Y2, Q9WVG9
Msl3
Recombinant protein encompassing a sequence within the center region of human MSL3L1. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.