Learn More
MSLN Monoclonal Antibody (OTI8H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA805262S
Encompass_Preferred
 View more versions of this product
Product Code. 17555113
  • € 242.00 / 30µL
Description
MSLN Monoclonal antibody specifically detects MSLN in Human samples. It is validated for Western Blot
Specifications
Specifications
MSLN
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13421
Mouse
Affinity chromatography
RUO
10232
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI8H7
Unconjugated
MSLN
CAK1; CAK1 antigen; megakaryocyte potentiating factor; Megakaryocyte-potentiating factor; Mes; Mesothelin; Mesothelin, cleaved form; MPF; MSLN; Pre-pro-megakaryocyte-potentiating factor; SMR; SMRP; soluble MPF mesothelin related protein
Human recombinant protein fragment corresponding to amino acids 270-493 of human MSLN produced in E.coli.
30 μL