missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSMB Monoclonal Antibody (OTI10F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803523S
This item is not returnable.
View return policy
Description
MSMB Monoclonal antibody specifically detects MSMB in Human samples. It is validated for Western BlotSpecifications
| MSMB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08118 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4477 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI10F4 | |
| Unconjugated | |
| MSMB | |
| beta-inhibin; beta-microseminoprotein; beta-MSP; HPC 13; HPC13; IGBF; immunoglobulin binding factor; Immunoglobulin-binding factor; microseminoprotein beta; microseminoprotein, beta; microseminoprotein, beta-; Msmb; MSP; MSPB; PIP; PN 44; PN44; Prostate secreted seminal plasma protein; prostate secretory protein of 94 amino acids; prostate secretory protein PSP94; prostatic inhibin protein; PRPS; PRSP; PSP; PSP 57; PSP57; PSP94; PSP-94; Seminal plasma beta-inhibin | |
| Purified prostate specific antigen (MSMB) from human seminal fluid | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction