missing translation for 'onlineSavingsMsg'
Learn More

MSMB Monoclonal Antibody (OTI2C3), TrueMAB™, OriGene

Product Code. p-200017104
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17512643 30 μL 30µL
17592633 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17512643 Supplier Origene Technologies Supplier No. TA803507S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MSMB Monoclonal antibody specifically detects MSMB in Human samples. It is validated for Western Blot

Specifications

Antigen MSMB
Applications Western Blot
Classification Monoclonal
Clone OTI2C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MSMB
Gene Accession No. P08118
Gene Alias beta-inhibin; beta-microseminoprotein; beta-MSP; HPC 13; HPC13; IGBF; immunoglobulin binding factor; Immunoglobulin-binding factor; microseminoprotein beta; microseminoprotein, beta; microseminoprotein, beta-; Msmb; MSP; MSPB; PIP; PN 44; PN44; Prostate secreted seminal plasma protein; prostate secretory protein of 94 amino acids; prostate secretory protein PSP94; prostatic inhibin protein; PRPS; PRSP; PSP; PSP 57; PSP57; PSP94; PSP-94; Seminal plasma beta-inhibin
Host Species Mouse
Immunogen Purified prostate specific antigen (MSMB) from human seminal fluid
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4477
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.