Learn More
MSMB Monoclonal Antibody (OTI8G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803517S
Encompass_Preferred
 View more versions of this product
Product Code. 17512673
  • € 242.00 / 30µL
Description
MSMB Monoclonal antibody specifically detects MSMB in Human samples. It is validated for Western Blot
Specifications
Specifications
MSMB
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P08118
Mouse
Affinity chromatography
RUO
4477
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI8G8
Unconjugated
MSMB
beta-inhibin; beta-microseminoprotein; beta-MSP; HPC 13; HPC13; IGBF; immunoglobulin binding factor; Immunoglobulin-binding factor; microseminoprotein beta; microseminoprotein, beta; microseminoprotein, beta-; Msmb; MSP; MSPB; PIP; PN 44; PN44; Prostate secreted seminal plasma protein; prostate secretory protein of 94 amino acids; prostate secretory protein PSP94; prostatic inhibin protein; PRPS; PRSP; PSP; PSP 57; PSP57; PSP94; PSP-94; Seminal plasma beta-inhibin
Purified prostate specific antigen (MSMB) from human seminal fluid