missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEKT2 Mouse anti-Human, Clone: OTI1E11, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809970
This item is not returnable.
View return policy
Description
Structural component of ciliary and flagellar microtubules. Plays a key role in the assembly or attachment of the inner dynein arm to microtubules in sperm flagella and tracheal cilia. Forms filamentous polymers in the walls of ciliary and flagellar microtubules.Specifications
| TEKT2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UIF3 | |
| TEKT2 | |
| Human recombit protein fragment corresponding to amino acids 1-244 of human TEKT2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1E11 | |
| Unconjugated | |
| TEKT2 | |
| TEKTB1, TEKTIN-T, h-tektin-t | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27285 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction