Learn More
MSMB Mouse anti-Human, Clone: OTI2A3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF803508
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MSMB protein is a member of the immunoglobulin binding factor family. It is synthesized by the epithelial cells of the prostate gland and secreted into the seminal plasma. This protein has inhibin-like activity. It may have a role as an autocrine paracrine factor in uterine, breast and other female reproductive tissues. The expression of the encoded protein is found to be decreased in prostate cancer. Two alternatively spliced transcript variants encoding different isoforms are described for this gene. The use of alternate polyadenylation sites has been found for this gene.Specifications
| MSMB | |
| Monoclonal | |
| Unconjugated | |
| MSMB | |
| HPC13, IGBF, MSP, MSPB, PN44, PRPS, PSP, PSP-94, PSP57, PSP94 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4477 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2A3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P08118 | |
| MSMB | |
| Purified prostate specific antigen (MSMB) from human seminal fluid | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.