Learn More
Novus Biologicals™ MSPR/Ron Beta Antibody (Aveo anti-RON) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28436-100ug
 View more versions of this product
Product Code. 30047443
  • € 338.00 / 100µg
Description
MSPR/Ron Beta Monoclonal antibody specifically detects MSPR/Ron Beta in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSPR/Ron Beta
Monoclonal
Unconjugated
Q04912
Human
Protein A purified
RUO
4486
Human
Purified
Flow Cytometry, ELISA, Functional Assay
Aveo anti-RON
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD136, CDw136, Macrophage-stimulating protein receptor beta chain, MSP receptor, NPCA3, p185-Ron, Protein-tyrosine kinase 8, PTK8, Ron
MSPR/ RON/ CD136
100 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.