missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSPR/Ron Beta Antibody (narnatumab), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28640CL1
This item is not returnable.
View return policy
Description
MSPR/Ron Beta Monoclonal antibody specifically detects MSPR/Ron Beta in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| MSPR/Ron Beta | |
| Monoclonal | |
| CoraFluor 1 | |
| CD136, CDw136, Macrophage-stimulating protein receptor beta chain, MSP receptor, NPCA3, p185-Ron, Protein-tyrosine kinase 8, PTK8, Ron | |
| MSPR/ RON/ CD136 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| narnatumab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| 4486 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction