missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSRA Polyclonal Antibody
Description
MSRA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Methionine sulfoxide reductase (MsrA) reduces methionine sulfoxide (MetO) residues in proteins and free MetO to Methionine (Met). The catalytic activity of MsrA is dependent of bound metal and cofactors but it requires reducing equivalents from either DTT or a thioredoxin-regenerating system. MsrA plays an essential role in protecting cells against oxidative damage. The substrates of MsrA include calmodulin, HIV protease and a 1-proteinase-inhibitor. Recent studies indicate that there is a connection between MsrA and Alzheimer's disease in mammals.
Specifications
Specifications
| Antigen | MSRA |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | Msra |
| Gene Accession No. | Q923M1, Q9UJ68 |
| Gene Alias | 2310045J23Rik; 6530413P12Rik; cytosolic methionine-S-sulfoxide reductase; methionine sulfoxide reductase A; mitochondrial peptide methionine sulfoxide reductase; Msra; MSR-A; peptide met (O) reductase; peptide Met(O) reductase; peptide methionine sulfoxide reductase; peptide-methionine (S)-S-oxide reductase; PMSR; protein-methionine-S-oxide reductase |
| Gene Symbols | Msra |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?