missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MSRA Polyclonal Antibody
GREENER_CHOICE

Product Code. 17780684
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17780684 200 μL 200µL
17723164 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17780684 Supplier Invitrogen™ Supplier No. PA5119282

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MSRA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Methionine sulfoxide reductase (MsrA) reduces methionine sulfoxide (MetO) residues in proteins and free MetO to Methionine (Met). The catalytic activity of MsrA is dependent of bound metal and cofactors but it requires reducing equivalents from either DTT or a thioredoxin-regenerating system. MsrA plays an essential role in protecting cells against oxidative damage. The substrates of MsrA include calmodulin, HIV protease and a 1-proteinase-inhibitor. Recent studies indicate that there is a connection between MsrA and Alzheimer's disease in mammals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSRA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Msra
Gene Accession No. Q923M1, Q9UJ68
Gene Alias 2310045J23Rik; 6530413P12Rik; cytosolic methionine-S-sulfoxide reductase; methionine sulfoxide reductase A; mitochondrial peptide methionine sulfoxide reductase; Msra; MSR-A; peptide met (O) reductase; peptide Met(O) reductase; peptide methionine sulfoxide reductase; peptide-methionine (S)-S-oxide reductase; PMSR; protein-methionine-S-oxide reductase
Gene Symbols Msra
Host Species Rabbit
Immunogen Recombinant protein Methionine Sulfoxide Reductase A. The antigen corresponds to amino acid range 27-235 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100152960, 29447, 4482
Target Species Human, Rat, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.