missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSRB3 Antibody (OTI2A2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72793UV
This item is not returnable.
View return policy
Description
MSRB3 Monoclonal specifically detects MSRB3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MSRB3 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MSRB3 | |
| Recombinant protein fragment corresponding to amino acids 33-192 of human MSRB3 (NP_932346) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2A2 | |
| Western Blot, Immunohistochemistry | |
| DFNB74, DKFZp686C1178, EC 1.8.4.-, EC 1.8.4.12, FLJ36866, methionine sulfoxide reductase B3, methionine-R-sulfoxide reductase B3, methionine-R-sulfoxide reductase B3, mitochondrial, MsrB3 | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 253827.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction