missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSS4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 197.00 - € 470.00
Specifications
| Antigen | MSS4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633841
|
Novus Biologicals
NBP2-93502-0.02ml |
0.02 mL |
€ 197.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620141
|
Novus Biologicals
NBP2-93502-0.1ml |
0.1 mL |
€ 470.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSS4 Polyclonal antibody specifically detects MSS4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MSS4 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 8395 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.1, MSS4Protein STM-7, Phosphatidylinositol-4-phosphate 5-kinase type I beta, phosphatidylinositol-4-phosphate 5-kinase type-1 beta, phosphatidylinositol-4-phosphate 5-kinase, type I, beta, PIP5K1-beta, PIP5KIbeta, PtdIns(4)P-5-kinase 1 beta, STM7EC 2.7.1.68, Type I phosphatidylinositol-4-phosphate 5-kinase beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 190-470 of human PIP5K1B (NP_003549.1). NVLPRSMRMHFTYDLKGSTYKRRASRKEREKSNPTFKDLDFLQDMHEGLYFDTETYNALMKTLQRDCRVLESFKIMDYSLLLGIHFLDHSLKEKEEETPQNVPDAKRTGMQKVLYSTAMESIQGPGKSGDGIITENPDTMGGIPAKSHRGEKLLLFMGIIDILQSYRLMKKLEHSWKALVYDGDTVSVHRPSFYADRFLKFMNSRVFKKIQALKASPSKKRCNSIAALKATSQEIVSSISQEWKDEKRDLLTEGQSFSSLDEEALGSRHRPDLVPSTPSLF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title