missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSS4 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97690IR
This item is not returnable.
View return policy
Description
MSS4 Polyclonal specifically detects MSS4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MSS4 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.7.1, MSS4Protein STM-7, Phosphatidylinositol-4-phosphate 5-kinase type I beta, phosphatidylinositol-4-phosphate 5-kinase type-1 beta, phosphatidylinositol-4-phosphate 5-kinase, type I, beta, PIP5K1-beta, PIP5KIbeta, PtdIns(4)P-5-kinase 1 beta, STM7EC 2.7.1.68, Type I phosphatidylinositol-4-phosphate 5-kinase beta | |
| Produced in rabbits immunized with E. coli-derived Human MSS4 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 8395 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction