missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MST1/STK4 Antibody (06), DyLight 405 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06387V
This item is not returnable.
View return policy
Description
MST1/STK4 Monoclonal antibody specifically detects MST1/STK4 in Human samples. It is validated for ELISA, ELISA, ELISA
Specifications
| MST1/STK4 | |
| Monoclonal | |
| DyLight 405 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| ELISA | |
| 06 | |
| 50mM Sodium Borate | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MST1/STK4 (Q13043-1; Glu2-Phe487). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction