missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MST1R, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB31654.100uL
This item is not returnable.
View return policy
Specifications
| MST1R | |
| Polyclonal | |
| ELISA (1:40000) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user. | |
| macrophage stimulating 1 receptor (c-met-related tyrosine kinase) | |
| MST1R | |
| A synthetic peptide corresponding to amino acids 1330-1410 of human MST1R (phospho S1394). | |
| RUO | |
| 4486 | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| In PBS (50% glycerol, 0.5% BSA and 0.02% sodium azide). | |
| CD136/CDw136/PTK8/RON | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of MST1R protein only when phosphorylated at S1394. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction