missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MST2/STK3 Monoclonal antibody specifically detects MST2/STK3 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | MST2/STK3 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 3C3 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA |
| Formulation | PBS, pH 7.4, 150mM NaCl and 50% glycerol |
| Gene Alias | EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, MST-2, MST2EC 2.7.11.1, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from Human MST2/STK3 [UniProt Q13188] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?