missing translation for 'onlineSavingsMsg'
Learn More

MST2/STK3 Antibody (3C3), Novus Biologicals™

Product Code. p-200022363 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
50 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30036617 50 μL 50µL
30038788 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30036617 Supplier Novus Biologicals Supplier No. NBP32629850ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MST2/STK3 Monoclonal antibody specifically detects MST2/STK3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MST2/STK3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3C3
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, MST-2, MST2EC 2.7.11.1, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human MST2/STK3 [UniProt Q13188]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 6788
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.