missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MST2/STK3 Antibody (OTI4G10), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72794AF405
This item is not returnable.
View return policy
Description
MST2/STK3 Monoclonal specifically detects MST2/STK3 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MST2/STK3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| STK3 | |
| Full-length protein expressed in 293T cell transfected with human MST2/STK3 expression vector. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI4G10 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, FLJ90748, KRS1, Mammalian STE20-like protein kinase 2, MST-2, MST2EC 2.7.11.1, serine/threonine kinase 3, serine/threonine kinase 3 (STE20 homolog, yeast), serine/threonine kinase 3 (Ste20, yeast homolog), serine/threonine-protein kinase 3, Serine/threonine-protein kinase Krs-1, STE20-like kinase MST2 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 6788.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction