Learn More
Eurofins DiscoverX MST4 Protein, Active
Description
MST4 is a member of the GCK group III family of kinases, which are a subset of the Ste20-like kinases. This protein kinase contains an amino-terminal kinase domain, and a carboxy-terminal regulatory domain that mediates homodimerization. The protein kinase localizes to the Golgi apparatus and is specifically activated by binding to the Golgi matrix protein GM130. It is also cleaved by caspase-3 in vitro, and may function in the apoptotic pathway.
GenBank NM_016542
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 38 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥69% |
| Protein | MST4, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.