Learn More
Invitrogen™ MSX1 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518284
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
MSX1 is a member of the muscle segment homeobox gene family. It functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. MSX1 may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in the gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal dominant hypodontia.
Specifications
| MSX1 | |
| Polyclonal | |
| Unconjugated | |
| MSX1 | |
| AA675338; AI324650; ECTD3; homeo box, msh-like 1; homeobox 7; homeobox protein Hox-7; Homeobox protein MSX-1; homeobox, msh-like 1; Hox7; Hox-7; Hox7.1; hox-7.1; HYD1; msh; msh homeo box 1; msh homeobox 1; msh homeobox 1-like protein; msh homeobox homolog 1; Msx1; muscle-segment homeobox; STHAG1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4487 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P28360 | |
| MSX1 | |
| Synthetic peptide sequence (TSLPLGVKVEDS) corresponding to the N-terminus amino acids of MSX1 (aa 8-20). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |