missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MT-ND1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140324
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PFDLAEGESE LVSGFNIEYA AG Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 1 (ND1) binds rotenone and rotenone analogs and might be involved in electron transfer to ubiquinone. Mutations in the ND1 gene may be implicated in several disorders, including Leber hereditary optic neuropathy, Alzheimer's disease and Parkinson disease.
Specifications
| MT-ND1 | |
| Polyclonal | |
| Unconjugated | |
| mt-Nd1 | |
| mitochondrially encoded NADH dehydrogenase 1; Mtnd1; MT-ND1; NADH dehydrogenase; NADH dehydrogenase subunit 1; NADH1; NADH-ubiquinone oxidoreductase chain 1; Nd1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4535 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P03886 | |
| mt-Nd1 | |
| Recombinant protein corresponding to Human MT-ND1. Recombinant protein control fragment (Product #RP-109116). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction