missing translation for 'onlineSavingsMsg'
Learn More

MT-ND3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15971705
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15971705 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15971705 Supplier Invitrogen Supplier No. PA575180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MT-ND3 Polyclonal Antibody

MT-ND3 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ND3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MT-ND3
Gene Accession No. P03899
Gene Alias 1.6.5.3; mitochondrially encoded NADH dehydrogenase 3; Mtnd3; mt-Nd3; NADH dehydrogenase subunit 3; NADH3; NADH-ubiquinone oxidoreductase chain 3; ND3
Gene Symbols MT-ND3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 21-34 of Mouse MT-ND3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17718
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.