missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ND4L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | MT-ND4L |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18251103
|
Novus Biologicals
NBP2-58026 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677848
|
Novus Biologicals
NBP2-58026-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MT-ND4L Polyclonal specifically detects MT-ND4L in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| MT-ND4L | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 4539 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LLVSISNTYGLDYVHNLNLLQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Complex I ND4L Subunit, EC 1.6.5.3, Mitochondrially Encoded NADH Dehydrogenase 4L, Mitochondrially Encoded NADH:Ubiquinone MTND4L, NADH Dehydrogenase 4L, NADH Dehydrogenase, Subunit 4L (Complex I), NADH4L, NADH-Ubiquinone Oxidoreductase Chain 4L, ND4L, Oxidoreductase Core Subunit 4L | |
| MT-ND4L | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title