missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MT-ND5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122075
This item is not returnable.
View return policy
Description
MT-ND5 Polyclonal Antibody for Western Blot, IHC (P)
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 5 (ND5) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encoding for ND5 are implicated in Mitochondrial neurogastrointestinal encephalomyopathy, an autosomal recessive multisystem disorder that results in dramatically elevated levels of circulating thymidine and deoxyuridine.
Specifications
| MT-ND5 | |
| Polyclonal | |
| Unconjugated | |
| MT-ND5 | |
| mitochondrially encoded NADH dehydrogenase 5; MTND5; mt-Nd5; NADH Dehydrogenase 5; NADH dehydrogenase subunit 5; NADH5; NADH-ubiquinone oxidoreductase chain 5; ND5 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 17721, 26202, 4540 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P03915, P03921, P11661 | |
| MT-ND5, ND5 | |
| Recombinant protein NADH Dehydrogenase 5. The antigen corresponds to amino acid range Arg424-Pro482 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction