missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MT-ND5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16529781
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16529781 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16529781 Supplier Invitrogen™ Supplier No. PA596945

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: Mouse heart, Mouse brain, Rat brain; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein Immunogen sequence: IALELNNLTM KLSMNKANPY SSFSTLLGFF PSIIHRITPM KSLNLSLKTS LTLLDLIWLE KTIPKSTSTL HTNMTTLTTN QKGLIKLYFM SFLINIILII ILYSINLE.

A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 5 (ND5) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encoding for ND5 are implicated in Mitochondrial neurogastrointestinal encephalomyopathy, an autosomal recessive multisystem disorder that results in dramatically elevated levels of circulating thymidine and deoxyuridine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ND5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MT-ND5
Gene Accession No. P03915, P03921, P11661
Gene Alias mitochondrially encoded NADH dehydrogenase 5; MTND5; mt-Nd5; NADH Dehydrogenase 5; NADH dehydrogenase subunit 5; NADH5; NADH-ubiquinone oxidoreductase chain 5; ND5
Gene Symbols MT-ND5, ND5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 500-607 of mouse MT-ND5 (NP_9043381).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17721, 26202, 4540
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.