missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MT-ND6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16650405 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16650405 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16650405 Supplier Invitrogen™ Supplier No. PA5103954

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MT-ND6.

Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ND6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene mt-Nd6
Gene Accession No. P03923, P03925, P03926
Gene Alias mitochondrially encoded NADH dehydrogenase 6; MTND6; MT-ND6; NADH dehydrogenase subunit 6; NADH6; NADH-ubiquinone oxidoreductase chain 6; ND6
Gene Symbols mt-Nd6, ND6
Host Species Rabbit
Immunogen A synthesized peptide derived from human MT-ND6(Accession P03923), corresponding to amino acid residues M54-L104.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17722, 26203, 4541
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.