missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTA2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56483F
This item is not returnable.
View return policy
Description
MTA2 Polyclonal specifically detects MTA2 in Human, Primate samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MTA2 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| DKFZp686F2281, metastasis associated 1 family, member 2, metastasis -associated gene 1-like 1, metastasis associated gene family, member 2, Metastasis-associated 1-like 1, metastasis-associated protein 2, metastasis-associated protein MTA2, MTA1-L1 protein, MTA1L1MTA1-L1, p53 target protein in deacetylase complex, PID | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MTA2 | |
| A portion of amino acids 650-700 of human MTA2 was used as the immunogen (NP_004730). | |
| 0.1 mL | |
| Cancer, Chromatin Research, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction