missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTA2 Recombinant Rabbit Monoclonal Antibody (9A5Y7)

MTA2 Recombinant Rabbit Monoclonal Antibody (ARC1056)
Brand: Invitrogen™ MA535547
This item is not returnable.
View return policy
Description
MTA2 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a protein that has been identified as a component of NuRD, a nucleosome remodeling deacetylase complex identified in the nucleus of human cells. It shows a very broad expression pattern and is strongly expressed in many tissues. It may represent one member of a small gene family that encode different but related proteins involved either directly or indirectly in transcriptional regulation. Their indirect effects on transcriptional regulation may include chromatin remodeling. It is closely related to another member of this family, a protein that has been correlated with the metastatic potential of certain carcinomas. These two proteins are so closely related that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. One of the proteins known to be a target protein for this gene product is p53. Deacetylation of p53 is correlated with a loss of growth inhibition in transformed cells supporting a connection between these gene family members and metastasis.
Specifications
| MTA2 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O94776 | |
| Mta2 | |
| A synthetic peptide corresponding to a sequence within amino acids 569-668 of human MTA2 (O94776). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ChIP Assay, ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 9A5Y7 | |
| Unconjugated | |
| Mta2 | |
| AW550797; DKFZp686F2281; Mata1l1; metastais-associated gene family, member 2; metastasis associated 1 family member 2; metastasis associated 1 family, member 2; metastasis associated 1-like 1; metastasis -associated gene 1-like 1; metastasis associated gene family, member 2; metastasis-associated 1-like 1; metastasis-associated gene family, member 2; metastasis-associated protein 2; metastasis-associated protein MTA2; mmta2; Mta1l1; MTA1-L1 protein; Mta2; p53 target protein in deacetylase complex; PID | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9219 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction