Learn More
MTA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14682-1-AP-150UL
This item is not returnable.
View return policy
Description
Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.Specifications
| MTA3 | |
| Polyclonal | |
| Unconjugated | |
| MTA3 | |
| KIAA1266, MTA3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100362346, 116871, 57504 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q924K8, Q9BTC8 | |
| MTA3 | |
| MTA3 Fusion Protein Ag6400 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |