missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTBP Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98002CL1
This item is not returnable.
View return policy
Description
MTBP Polyclonal antibody specifically detects MTBP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MTBP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| hMTBP, MDM2 (mouse double minute 2)-binding protein, 104kD, Mdm2, transformed 3T3 cell double minute 2, p53 binding protein (mouse) bindingprotein, 104kDa, mdm2-binding protein, MDM2BP | |
| Produced in rabbits immunized with E. coli-derived Human MTBP fragment. | |
| 0.1 mL | |
| Cancer, DNA Repair | |
| 27085 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction