missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTCH1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1303
This item is not returnable.
View return policy
Description
MTCH1 Polyclonal specifically detects MTCH1 in Human, Mouse, Rat, Canine samples. It is validated for Peptide ELISA.
Specifications
| MTCH1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| cell proliferation-inducing protein 60, CGI-64, MGC131998, mitochondrial carrier homolog 1, mitochondrial carrier homolog 1 (C. elegans), Presenilin-associated protein, PSAPPIG60 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:16000 | |
| NP_055156. | |
| MTCH1 | |
| Peptide with sequence FRRVSSGSCFALE corresponding to C-Terminus according to NP_055156.1. | |
| 0.1 mg | |
| Apoptosis, Lipid and Metabolism | |
| 23787 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction