missing translation for 'onlineSavingsMsg'
Learn More

MTCH2 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18619831 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18619831 0.1 mL 0.10mL
18668761 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18619831 Supplier Novus Biologicals Supplier No. NBP2941850.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MTCH2 Polyclonal antibody specifically detects MTCH2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTCH2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias 2310034D24Rik, Met-induced mitochondrial protein, MIMP, mitochondrial carrier 2, mitochondrial carrier homolog 2, mitochondrial carrier homolog 2 (C. elegans)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-175 of human MTCH2 (NP_055157.1). VGYEPLPPTIGRNIFGRQVCQLPGLFSYAQHIASIDGRRGLFTGLTPRLCSGVLGTVVHGKVLQHYQESDKGEELGPGNVQKEVSSSFDHVIKETTREMIARSAATLITHPFHVITLRSMVQFIGRESKYCGLCDSIITIYREEGI
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23788
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.