missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTDH Monoclonal Antibody (OTI1E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
POA
Specifications
| Antigen | MTDH |
|---|---|
| Clone | OTI1E7 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17512943
|
Origene Technologies
TA803707S |
30 μL |
POA
30µL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17592933
|
Origene Technologies
TA803707 |
100 μL |
POA
100µL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MTDH Monoclonal antibody specifically detects MTDH in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MTDH | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| IgG1 | |
| -20° C, Avoid Freeze/Thaw Cycles |
| OTI1E7 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Primary |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.