Learn More
MTDH Mouse anti-Human, Clone: OTI4C5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF803793
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Metadherin (Metastasis adhesion protein), also known as MTDH, LYsine-RIch CEACAM1 co-isolated (LYRIC), is a novel protein that localizes with the tight junction proteins ZO-1 and occludin in polarized epithelial cells. At the tight junction, it acts not as a structural component, but is rather recruited during the maturation of the tight junction complex. Metadherin is overexpressed in breast cancer tissue and breast tumor xenografts, while much lower levels are expressed in normal breast tissue. Metadherin binds to lung vasculature, one of the four common sites of breast cancer metastasis, through a C-terminal segment in the extracellular domain; blocking this lung-homing domain with antibodies or inhibiting metadherin with siRNA has been reported to inhibit breast cancer metastasis.Specifications
| MTDH | |
| Monoclonal | |
| Unconjugated | |
| MTDH | |
| 3D3, AEG-1, AEG1, LYRIC, LYRIC/3D3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 92140 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4C5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q86UE4 | |
| MTDH | |
| Human recombit protein fragment corresponding to amino acids 70-582 of human MTDH produced in SF9 cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.