Learn More
MTERFD1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-81740CL1
Product Code. 30153579
  • € 965.00 / 0.10mL
Description
MTERFD1 Polyclonal antibody specifically detects MTERFD1 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MTERFD1
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse
Purified
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
CoraFluor 1
CGI-12, FLJ10939, Mitochondrial transcription termination factor 3, MTERF domain containing 1, mTERF domain-containing protein 1, mitochondrial, mTERF3
MTERFD1 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MTERFD1 . The immunogen is located within the last 50 amino acids of MTERFD1. Amino Acid Squence: LGRAQYDPAKPNYISLD
0.1 mL
Autophagy, Cell Biology, Endocrinology
51001
Store at 4°C in the dark. Do not freeze.