missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERFD1 Polyclonal Antibody

MTERFD1 Polyclonal Antibody
Brand: Invitrogen PA598630
This item is not returnable.
View return policy
Description
MTERFD1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| MTERFD1 | |
| Polyclonal | |
| Unconjugated | |
| MTERF3 | |
| 2410017I18Rik; Cgi12; CGI-12; Mitochondrial transcription termination factor 3; MTERF domain containing 1; mTERF domain-containing protein 1, mitochondrial; Mterf3; MTERFD1; transcription termination factor 3, mitochondrial | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51001 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.65 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96E29 | |
| MTERF3 | |
| Recombinant Human Transcription termination factor 3, mitochondrial protein (69-300AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction