missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERFD2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81966AF488
This item is not returnable.
View return policy
Description
MTERFD2 Polyclonal antibody specifically detects MTERFD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MTERFD2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 130916 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| FLJ16261, MGC61716, MTERF domain containing 2, mTERF domain-containing protein 2 | |
| MTERFD2 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MTERFD2 . The immunogen is located within amino acids 260 - 310 of MTERFD2. Amino Acid Squence: LERLGRYQTPDKKGQTQI | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction