missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 183.00 - € 322.00
Specifications
| Antigen | MTF1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18674299
|
Novus Biologicals
NBP3-12562-100ul |
100 μg |
€ 322.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641909
|
Novus Biologicals
NBP3-12562-50ul |
50 μg |
€ 183.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTF1 Polyclonal antibody specifically detects MTF1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MTF1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| metal regulatory transcription factor 1, metal-regulatory transcription factor 1, metal-responsive transcription factor 1, MGC23036, MRE-binding transcription factor, MRE-binding transcription factor-1, MTF-1, Transcription factor MTF-1, zinc regulatory factor, ZRF | |
| Produced in rabbits immunized with E. coli-derived Human MTF1 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 4520 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title