Learn More
MTF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555916
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EHSPDNNIIY FEAEEDELTP DDKMLRFVDK NGLVPSSSGT VYDRTTVLIE QDPGTLEDED DDGQCG Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
MTF1 Polyclonal antibody specifically detects MTF1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
MTF1 | |
Polyclonal | |
Unconjugated | |
MTF1 | |
metal regulatory transcription factor 1; metal response element binding transcription factor 1; metalloregulatory transcription factor; metal-regulatory transcription factor 1; metal-responsive transcription factor 1; MRE-binding transcription factor; MRE-binding transcription factor-1; Mtf1; MTF-1; Thyls; Transcription factor MTF-1; zinc regulatory factor; ZRF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4520 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14872 | |
MTF1 | |
Recombinant protein corresponding to Human MTF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only