missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTFMT Antibody (OTI2A2), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72803AF594
This item is not returnable.
View return policy
Description
MTFMT Monoclonal specifically detects MTFMT in Human samples. It is validated for Western Blot.
Specifications
| MTFMT | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MTFMT | |
| Recombinant protein fragment corresponding to amino acids 175-389 of human MTFMT (NP_640335) produced in E.coli. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI2A2 | |
| Western Blot | |
| EC 2.1.2.9, FMT, FMT1methionyl-tRNA formyltransferase, mitochondrial, mitochondrial methionyl-tRNA formyltransferase, MtFMT | |
| Mouse | |
| 0.1 mL | |
| 123263 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction