missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTFMT Antibody (OTI2A2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46028
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
MTFMT Monoclonal specifically detects MTFMT in Human samples. It is validated for Western Blot.
Specifications
| MTFMT | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| EC 2.1.2.9, FMT, FMT1methionyl-tRNA formyltransferase, mitochondrial, mitochondrial methionyl-tRNA formyltransferase, MtFMT | |
| Mouse | |
| 43.7 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 123263 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI2A2 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| MTFMT | |
| Human recombinant protein fragment corresponding to amino acids 175-389 of human MTFMT(NP_640335) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction