missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MTFR1L Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA555516

Product Code. 15703441

  • € 560.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SEERLLALKK PALPALSRTT ELQDELSHLR SQIAKIVAAD AASASLTPDF LSPGSSNVSS PLPCFGSSFH STTS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 99%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MTFR1L
Polyclonal
Unconjugated
MTFR1L
FAM54B; family with sequence similarity 54 member B; family with sequence similarity 54, member B; HYST1888; mitochondrial fission regulator 1 like; mitochondrial fission regulator 1-like; MST116; MSTP116; MTFR1L; protein FAM54B
Rabbit
Antigen Affinity Chromatography
RUO
56181
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H019
MTFR1L
Recombinant protein corresponding to Human MTFR1L. Recombinant protein control fragment (Product #RP-94155).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.