missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MTG1 (Human) Recombinant Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
- Sequence: MAKGLKKMQSSLKLVDCIIEVHDARIPLSGRNPLFQETLGLKPHLLVLNKMDLADLTEQQKIMQHLEGEGLKNVIFTNCVKDENVKQIIPMVTELIGRSHRYHRKENLEYCIMVIGVPNVGKSSLINSLRRQHLRKGKATRVGGEPGITRAVMSKIQVSERPLMFLLDTPGVLAPRIESVETGLKLALCGTVLDHLVGEETMADYLLYTLNKHQRFGYVQHYGLGSACDNVERVLKSVAVKLGKTQKVKVLTGTGNVNVIQPNYPAAARDFLQTFRRGLLGSVMLDLDVLRGHPPAETLP
Specifications
Specifications
| Accession Number | AAH35721.1 |
| Gene ID (Entrez) | 92170 |
| Name | mitochondrial GTPase 1 homolog (S. cerevisiae) |
| Preparation Method | Wheat germ expression system |
| Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | GTP, GTPBP7, RP11-108K14.2 |
| Gene Symbol | MTG1 |
| Species | Wheat Germ (in vitro) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction