missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTG2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103868
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GTPBP5.
Small G proteins, such as GTPBP5, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 ).
Specifications
| MTG2 | |
| Polyclonal | |
| Unconjugated | |
| MTG2 | |
| 1810011P19Rik; 2900056P18Rik; D2Bwg0647e; dJ1005F21.2; GTP binding protein 5; GTP binding protein 5 (putative); GTP-binding protein 5; GTP-binding protein 5 (putative); Gtpbp5; mitochondrial ribosome associated GTPase 2; mitochondrial ribosome-associated GTPase 2; Mtg2; OBGH1; protein obg homolog 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26164, 296462, 52856 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H4K7 | |
| MTG2 | |
| A synthesized peptide derived from human MTG2(Accession Q9H4K7), corresponding to amino acid residues C303-Q353. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction