missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTGR1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97748G
This item is not returnable.
View return policy
Description
MTGR1 Polyclonal antibody specifically detects MTGR1 in Human samples. It is validated for Western Blot
Specifications
| MTGR1 | |
| Polyclonal | |
| Western Blot | |
| core-binding factor, runt domain, alpha subunit 2; translocated to, 2, EHTDKFZp313F2116, ETO homolog on chromosome 20, ETO homologous on chromosome 20, MTG8-like protein, MTG8-related protein 1, MTGR1p85, myeloid translocation gene-related protein 1, Myeloid translocation-related protein 1, protein CBFA2T2, ZMYND3 | |
| The immunogen recognized by this antibody maps to a region between residue 554 and 604 of human Core-Binding Factor, Runt Domain, Alpha Subunit 2; translocated to, 2 using the numbering given in entry NP_005084.1 (GeneID 9139). | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 9139 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction