missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTGR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584458
This item is not returnable.
View return policy
Description
Immunogen sequence: MAKESGISLK EIQVLARQWK VGPEKRVPAM PGSPVEVKIQ SRSSPPTMPP L Highest antigen sequence indentity to the following orthologs: Rat - 55%, Mouse - 57%.
Myeloid tumor related gene (MTGR1) is found deleted in acute myeloid leukemia. Overexpression of MTGR1 can stimulate AML1-MTG8 to induce GCSF- dependent proliferation of L-G cells and to interfere with AML1- dependent transcription. AML1/MTG8 functions as a complex with MTGR1 and that the complex might be important in promoting leukemogenesis.
Specifications
| MTGR1 | |
| Polyclonal | |
| Unconjugated | |
| CBFA2T2 | |
| A430091M07; C330013D05Rik; CBFA2/RUNX1 partner transcriptional co-repressor 2; CBFA2/RUNX1 translocation partner 2; CBFA2T2; CBFA2T2 identified gene homolog; Cbfa2t2h; core-binding factor runt domain alpha subunit 2 translocated to 2 homolog; core-binding factor, runt domain, alpha subunit 2, translocated to, 2 (human); core-binding factor, runt domain, alpha subunit 2, translocated to, 2 homolog; core-binding factor, runt domain, alpha subunit 2; translocated to, 2; DKFZp313F2116; EHT; ETO homolog on chromosome 20; ETO homologous on chromosome 20; MTG8-like protein; MTG8-related protein 1; MTGR1; myeloid translocation gene-related protein 1; myeloid translocation-related protein 1; p85; Protein CBFA2T2; RP5-1137F22.1; translocated to, 2; ZMYND3; ZMYND3 CBFA2T2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9139 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43439 | |
| CBFA2T2 | |
| Recombinant protein corresponding to Human CBFA2T2. Recombinant protein control fragment (Product #RP-101124). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction