missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTH1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97131AF750
This item is not returnable.
View return policy
Description
MTH1 Polyclonal antibody specifically detects MTH1 in Human samples. It is validated for ELISA
Specifications
| MTH1 | |
| Polyclonal | |
| ELISA | |
| 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase, EC 3.6.1.-, MTH1Nucleoside diphosphate-linked moiety X motif 1, mutT human homolog 1,8-oxo-dGTPase, nucleoside diphosphate-linked moiety X-type motif 17,8-dihydro-8-oxoguanine triphosphatase, nudix (nucleoside diphosphate linked moiety X)-type motif 1,8-oxo-7,8-dihydroguanosine triphosphatase, Nudix motif 1 | |
| Produced in rabbits immunized with purified, recombinant Human MTH1 (NP_002443.3; Met1-Val156). | |
| 0.1 mL | |
| Base Excision Repair, Cancer, Chromatin Research, DNA Repair, Modulation of DNA Pools, Stem Cell Markers | |
| 4521 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction