missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFD1L Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68321-1-IG-150UL
This item is not returnable.
View return policy
Description
The MTHFD1L antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MTHFD1L. This antibody recognizes human, mouse, rat, rabbit antigen. The MTHFD1L antibody has been validated for the following applications: WB, IHC, FC (Intra), ELISA analysis.Specifications
| MTHFD1L | |
| Monoclonal | |
| 1000 μg/mL | |
| DKFZp586G1517; FLJ21145; FTHFSDC1; MTC1THFS; dJ292B18.2 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| 106 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), ELISA | |
| 1G1F2 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 25902 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction