missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTHFD1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100158
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MTHFD1L.
One-carbon substituted forms of tetrahydrofolate (THF) are involved in the de novo synthesis of purines and thymidylate and support cellular methylation reactions through the regeneration of methionine from homocysteine. MTHFD1L is an enzyme involved in THF synthesis in mitochondria (Christensen et al., 2005 [PubMed 15611115]).
Specifications
| MTHFD1L | |
| Polyclonal | |
| Unconjugated | |
| MTHFD1L | |
| 10-formyl-THF synthetase; 2410004L15Rik; AI647056; dJ292B18.2; Formyltetrahydrofolate synthetase; formyltetrahydrofolate synthetase domain containing 1; FTHFSDC1; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like; monofunctional C1-tetrahydrofolate synthase, mitochondrial; MTC1THFS; MTHFD1L | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25902, 270685, 361472 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3V3R1, Q6UB35 | |
| MTHFD1L | |
| A synthesized peptide derived from human MTHFD1L(Accession Q6UB35), corresponding to amino acid residues F942-F978. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction